ENA238710832 Enamine

1-(butane-1-sulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD08445148
  • Cas Number : 923155-53-7
  • Molecular Weight : 249.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43131
Availability details for
Quantity
  • Availability
  • Add to Cart