ENA238710837 Enamine

1-(butane-1-sulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08445030
  • Cas Number : 1160562-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43169
Availability details for
Quantity
  • Availability
  • Add to Cart