ENA238710846 Enamine

4-chloro-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD09837290
  • Cas Number : 444909-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43224
Availability details for
Quantity
  • Availability
  • Add to Cart