ENA238710854 Enamine

1-(2H-1,2,3,4-tetrazol-5-yl)ethan-1-amine

  • MDL number MFCD12912749
  • Cas Number : 33876-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 267-268
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43283
Availability details for
Quantity
  • Availability
  • Add to Cart