ENA238710861 Enamine

2-(3-nitrophenyl)imidazo[1,2-a]pyridine

  • MDL number MFCD00444901
  • Cas Number : 34658-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43323
Availability details for
Quantity
  • Availability
  • Add to Cart