ENA238710867 Enamine

1,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD10478930
  • Cas Number : 1015599-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43374

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43374
Availability details for
Quantity
  • Availability
  • Add to Cart