ENA238710873 Enamine

4-methyl-2-(4-nitrophenyl)-1,3-thiazole

  • MDL number MFCD12197012
  • Cas Number : 109029-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43415

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43415
Availability details for
Quantity
  • Availability
  • Add to Cart