ENA238710874 Enamine

2-(propan-2-yl)-5H,6H-[1,2,4]triazolo[1,5-c]quinazolin-5-one

  • MDL number MFCD11858073
  • Cas Number : 1146290-29-0
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43416
Availability details for
Quantity
  • Availability
  • Add to Cart