ENA238710880 Enamine

2-{4-[(3-aminophenyl)methoxy]phenyl}acetonitrile

  • MDL number MFCD09811014
  • Cas Number : 1016750-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-43438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43438
Availability details for
Quantity
  • Availability
  • Add to Cart