ENA238710885 Enamine

N-[(furan-2-yl)methyl]pyridin-4-amine

  • MDL number MFCD11121729
  • Cas Number : 880878-30-8
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43462
Availability details for
Quantity
  • Availability
  • Add to Cart