ENA238710887 Enamine

(1-methyl-1,2,3,4-tetrahydroquinolin-4-yl)methanamine

  • MDL number MFCD12912753
  • Cas Number : 646997-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43478
Availability details for
Quantity
  • Availability
  • Add to Cart