ENA238710888 Enamine

5-oxo-1H,5H-pyrazolo[1,5-a]quinazoline-3-carboxylic acid

  • MDL number MFCD12197014
  • Cas Number : 1181458-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43481
Availability details for
Quantity
  • Availability
  • Add to Cart