ENA238710890 Enamine

3-[(2-bromophenyl)methoxy]benzoic acid

  • MDL number MFCD08444844
  • Cas Number : 923809-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43491
Availability details for
Quantity
  • Availability
  • Add to Cart