ENA238710897 Enamine

N-[2-(dimethylamino)ethyl]-1-methylpiperidin-4-amine

  • MDL number MFCD03856661
  • Cas Number : 724757-63-5
  • Molecular Weight : 185.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43520

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-43520
pictograms GHS05,GHS07
hstatements H227,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart