ENA238710910 Enamine

2-(pyrrolidin-1-yl)acetonitrile

  • MDL number MFCD00014098
  • Cas Number : 29134-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43581

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-43581
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart