ENA238710917 Enamine

5-phenyl-4-(1-phenylethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD11982546
  • Cas Number : 1038324-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43610
Availability details for
Quantity
  • Availability
  • Add to Cart