ENA238710925 Enamine

3-(3-bromobenzamido)benzoic acid

  • MDL number MFCD00433570
  • Cas Number : 413574-81-9
  • Molecular Weight : 320.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43657
Availability details for
Quantity
  • Availability
  • Add to Cart