ENA238710929 Enamine

2-ethoxy-4-fluoroaniline

  • MDL number MFCD07801192
  • Cas Number : 178993-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43682
Availability details for
Quantity
  • Availability
  • Add to Cart