ENA238710933 Enamine

2-[(6-bromonaphthalen-2-yl)oxy]acetic acid

  • MDL number MFCD06260704
  • Cas Number : 141791-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43725
Availability details for
Quantity
  • Availability
  • Add to Cart