ENA238710934 Enamine

4-[1-(cyclopropylamino)ethyl]phenol

  • MDL number MFCD09047625
  • Cas Number : 926204-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43730

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43730
Availability details for
Quantity
  • Availability
  • Add to Cart