ENA238710937 Enamine

2-amino-4-methylpentanenitrile hydrochloride

  • MDL number MFCD11858127
  • Cas Number : 72177-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43747
Availability details for
Quantity
  • Availability
  • Add to Cart