ENA238710938 Enamine

ethyl 6-methyl-2-phenylpyrimidine-4-carboxylate

  • MDL number MFCD12197021
  • Cas Number : 1147675-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43750
Availability details for
Quantity
  • Availability
  • Add to Cart