ENA238710942 Enamine

1-[2-(4-ethoxyphenoxy)pyridin-4-yl]methanamine

  • MDL number MFCD09737215
  • Cas Number : 954273-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 78
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43761
Availability details for
Quantity
  • Availability
  • Add to Cart