ENA238710944 Enamine

1-{3-methoxy-4-[(pyridin-4-yl)methoxy]phenyl}ethan-1-amine

  • MDL number MFCD09733971
  • Cas Number : 953753-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-43764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43764
Availability details for
Quantity
  • Availability
  • Add to Cart