ENA238710946 Enamine

5-chloro-1-methyl-3-(propan-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11185414
  • Cas Number : 1043918-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43769
Availability details for
Quantity
  • Availability
  • Add to Cart