ENA238710953 Enamine

2-[(3-fluorophenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD05241569
  • Cas Number : 57978-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43795
Availability details for
Quantity
  • Availability
  • Add to Cart