ENA238710960 Enamine

4-(1,1,2,2,2-pentafluoroethoxy)aniline

  • MDL number MFCD10686955
  • Cas Number : 717-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43809

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43809
Availability details for
Quantity
  • Availability
  • Add to Cart