ENA238710966 Enamine

6-(2-bromo-4-fluorophenoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09730927
  • Cas Number : 953901-37-6
  • Molecular Weight : 312.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(2-bromo-4-fluorophenoxy)nicotinic acid
Inchi Code 1S/C12H7BrFNO3/c13-9-5-8(14)2-3-10(9)18-11-4-1-7(6-15-11)12(16)17/h1-6H,(H,16,17)
Inchi Key PGPHGIWPCDRYEY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43826
Availability details for
Quantity
  • Availability
  • Add to Cart