ENA238710971 Enamine

1,8-dioxo-1H,2H,3H,4H,8H-pyrido[1,2-a]piperazine-6-carboxylic acid

  • MDL number MFCD01104311
  • Cas Number : 5368-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43836
Availability details for
Quantity
  • Availability
  • Add to Cart