ENA238710974 Enamine

5-[(2E)-2-[(2-bromophenyl)methylidene]hydrazin-1-yl]-2-chlorobenzoic acid

  • MDL number MFCD02722692
  • Cas Number : 380215-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43841
Availability details for
Quantity
  • Availability
  • Add to Cart