ENA238710976 Enamine

(2-hydroxyphenyl)(1-phenyl-1H-pyrazol-4-yl)methanone phenylhydrazone

  • MDL number MFCD05263112
  • Cas Number : 74990-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08651

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08651
Availability details for
Quantity
  • Availability
  • Add to Cart