ENA238710977 Enamine

5,7-dichloro-2lambda4,1,3-benzothiadiazol-4-amine

  • MDL number MFCD06335070
  • Cas Number : 16407-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-08915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08915
Availability details for
Quantity
  • Availability
  • Add to Cart