ENA238710982 Enamine

1-{4-[(1-methyl-1H-imidazol-2-yl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD11182282
  • Cas Number : 1094382-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43866
Availability details for
Quantity
  • Availability
  • Add to Cart