ENA238710985 Enamine

1-(4-bromophenyl)-3-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00466111
  • Cas Number : 14580-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43871
Availability details for
Quantity
  • Availability
  • Add to Cart