ENA238710988 Enamine

[2-(3-chloropropoxy)ethyl]benzene

  • MDL number MFCD10695667
  • Cas Number : 286440-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43878
Availability details for
Quantity
  • Availability
  • Add to Cart