ENA238710989 Enamine

2-chloro-N-(2lambda4,1,3-benzothiadiazol-4-yl)acetamide

  • MDL number MFCD05263778
  • Cas Number : 175203-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09128
Availability details for
Quantity
  • Availability
  • Add to Cart