ENA238710990 Enamine

2lambda4,1,3-benzothiadiazole-4-sulfonyl chloride

  • MDL number MFCD04066702

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09343
Availability details for
Quantity
  • Availability
  • Add to Cart