ENA238710992 Enamine

1',2'-dihydrospiro[cyclohexane-1,3'-indole]-2',4-dione

  • MDL number MFCD12197039
  • Cas Number : 52140-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-200
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43893
Availability details for
Quantity
  • Availability
  • Add to Cart