ENA238710995 Enamine

methyl 2-methyl-5-nitropyridine-3-carboxylate

  • MDL number MFCD12197040
  • Cas Number : 936130-27-7
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43898
Availability details for
Quantity
  • Availability
  • Add to Cart