ENA238711013 Enamine

2-{[(2-chloro-6-fluorophenyl)methyl]amino}acetic acid

  • MDL number MFCD08443290
  • Cas Number : 926270-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43942
Availability details for
Quantity
  • Availability
  • Add to Cart