ENA238711017 Enamine

5,6-dimethyl-3-[(pyridin-2-yl)methyl]-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD04969630
  • Cas Number : 861239-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43950
Availability details for
Quantity
  • Availability
  • Add to Cart