ENA238711019 Enamine

N-[1-(pyridin-2-yl)ethyl]cyclopropanamine

  • MDL number MFCD09044472
  • Cas Number : 867008-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43957
Availability details for
Quantity
  • Availability
  • Add to Cart