ENA238711020 Enamine

6-(oxiran-2-yl)-2,3-dihydro-1,4-benzodioxine

  • MDL number MFCD09728031
  • Cas Number : 389124-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43961
Availability details for
Quantity
  • Availability
  • Add to Cart