ENA238711023 Enamine

2-(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD01114908
  • Cas Number : 374913-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43969
Availability details for
Quantity
  • Availability
  • Add to Cart