ENA238711025 Enamine

7-(piperazin-1-ylsulfonyl)-2,1,3-benzothiadiazol

  • MDL number MFCD06355956
  • Cas Number : 488122-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...183
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10869
Availability details for
Quantity
  • Availability
  • Add to Cart