ENA238711028 Enamine

2-(4-bromophenoxy)-2-phenylacetic acid

  • MDL number MFCD09705805
  • Cas Number : 938147-43-4
  • Molecular Weight : 307.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43979
Availability details for
Quantity
  • Availability
  • Add to Cart