ENA238711032 Enamine

3-[(2-phenoxyethoxy)methyl]aniline

  • MDL number MFCD09807261
  • Cas Number : 1016749-42-0
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43985
Availability details for
Quantity
  • Availability
  • Add to Cart