ENA238711036 Enamine

4-phenyl-N1-(phenylmethylidene)-1H-imidazole-1,2-diamine

  • Cas Number : 28734-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11070
Availability details for
Quantity
  • Availability
  • Add to Cart