ENA238711042 Enamine

2-[2-(3-fluorophenyl)acetamido]acetic acid

  • MDL number MFCD09807218
  • Cas Number : 1016527-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44017
Availability details for
Quantity
  • Availability
  • Add to Cart