ENA238711048 Enamine

N-cyclopropyl-4-hydroxybenzamide

  • MDL number MFCD11130694
  • Cas Number : 860298-71-1
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44030
Availability details for
Quantity
  • Availability
  • Add to Cart